This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03356081_m1
See other FAR1 GE Assays ›
Gene Symbol:
FAR1
Gene Name:
fatty acyl-CoA reductase 1
Gene Aliases:
MLSTD2
Chromosome Location:
Chr.5: 7710324 - 7793832 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03356081_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031179.1 NP_001026350.1 7 - 8 1007 76
XM_015286359.2 XP_015141845.1 7 - 8 922 76
XM_015286360.2 XP_015141846.1 7 - 8 924 76
XM_015286362.2 XP_015141848.1 7 - 8 923 76
XM_015286363.2 XP_015141849.1 7 - 8 924 76
GenBank mRNA AJ719512.1 - 7 - 8 1007 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ether lipid biosynthetic process
wax biosynthetic process
long-chain fatty-acyl-CoA metabolic process
glycerophospholipid biosynthetic process
oxidation-reduction process
fatty-acyl-CoA reductase (alcohol-forming) activity
alcohol-forming fatty acyl-CoA reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0