This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352435_m1
See other MFSD13A GE Assays ›
Gene Symbol:
MFSD13A
Gene Name:
major facilitator superfamily domain containing 13A
Gene Aliases:
TMEM180
Chromosome Location:
Chr.6: 24108244 - 24121177 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352435_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFSD13A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031232.1 NP_001026403.1 7 - 8 1233 68
XM_015288635.2 XP_015144121.1 7 - 8 1368 68
XM_015288636.2 XP_015144122.1 7 - 8 1266 68
XM_015288637.2 XP_015144123.1 7 - 8 1237 68
GenBank mRNA AJ720089.1 - 7 - 8 1233 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0