This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352403_mH
See other ACTA2 GE Assays ›
Gene Symbol:
ACTA2
Gene Name:
actin, alpha 2, smooth muscle, aorta
Gene Aliases:
asma
Chromosome Location:
Chr.6: 20062662 - 20075102 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352403_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031229.1 NP_001026400.1 2 - 3 250 95
XM_015288391.2 XP_015143877.1 3 - 4 338 95
XM_015288392.2 XP_015143878.1 3 - 4 321 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
mesenchyme migration
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0