This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352062_m1
See other MRPS7 GE Assays ›
Gene Symbol:
MRPS7
Gene Name:
mitochondrial ribosomal protein S7
Gene Aliases:
-
Chromosome Location:
Chr.18: 10717538 - 10723317 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352062_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPS7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031109.3 NP_001026280.2 5 - 6 607 60
XM_015295405.2 XP_015150891.1 5 - 6 588 60
XM_015295406.2 XP_015150892.1 5 - 6 731 60
XM_015295407.2 XP_015150893.1 5 - 6 489 60
XM_025141616.1 XP_024997384.1 5 - 6 629 60
GenBank mRNA AJ719294.1 - 5 - 6 598 60
BX934833.1 - 5 - 6 658 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
translation
mitochondrial translation
mRNA binding
structural constituent of ribosome
rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0