This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352007_m1
See other MYCN GE Assays ›
Gene Symbol:
MYCN
Gene Name:
v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
Gene Aliases:
-
Chromosome Location:
Chr.3: 99207625 - 99212868 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352007_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031091.2 NP_001026262.2 1 - 2 725 108
XM_015276044.2 XP_015131530.1 2 - 3 1119 108
XM_015276045.2 XP_015131531.1 2 - 3 1544 108
XM_015276046.2 XP_015131532.1 2 - 3 1232 108
XM_015276048.2 XP_015131534.1 2 - 3 1162 108
XM_015276049.2 XP_015131535.1 2 - 3 1086 108
GenBank mRNA X68074.1 - 80 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0