This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03351404_m1
See other SPPL2C GE Assays ›
Gene Symbol:
SPPL2C
Gene Name:
signal peptide peptidase like 2C
Gene Aliases:
SPPL2B
Chromosome Location:
Chr.28: 659696 - 694735 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03351404_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPPL2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030933.1 NP_001026104.1 4 - 5 550 109
XM_015299708.2 XP_015155194.1 4 - 5 473 109
GenBank mRNA AJ851767.1 - 4 - 5 550 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

aspartic protease

Gene Ontology Categories:

Function(s) Process(es)

membrane protein ectodomain proteolysis
membrane protein intracellular domain proteolysis
membrane protein proteolysis
regulation of immune response
aspartic endopeptidase activity, intramembrane cleaving
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0