This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03347559_m1
See other CHD1Z GE Assays ›
Gene Symbol:
CHD1Z
Gene Name:
chromodomain helicase DNA binding protein 1-Z
Gene Aliases:
CHD-Z, CHD1, CHDZ
Chromosome Location:
Chr.Z: 51359549 - 51407969 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03347559_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD1Z
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204941.1 NP_990272.1 5 - 6 661 76
GenBank mRNA AB465210.1 - 4 - 5 431 76
AF004397.1 - 5 - 6 661 76
CR354120.1 - 5 - 6 885 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0