This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03346982_m1
See other GAPDH GE Assays ›
Gene Symbol:
GAPDH
Gene Name:
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases:
KNC-NDS6
Chromosome Location:
Chr.1: 77619440 - 77623355 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03346982_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GAPDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204305.1 NP_989636.1 11 - 12 983 107
GenBank mRNA AF047874.1 - 11 - 12 983 107
AY466166.1 - 4 - 5 342 107
K01458.1 - 11 - 12 982 107
V00407.1 - 10 - 11 955 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Endogenous Controls Recommended Assay Gallus gallus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
nuclear membrane fusion
gluconeogenesis
glycolytic process
negative regulation of endopeptidase activity
negative regulation of translation
peptidyl-cysteine S-trans-nitrosylation
negative regulation of apoptotic process
positive regulation of cytokine secretion
protein stabilization
defense response to fungus
neuron apoptotic process
killing by host of symbiont cells
positive regulation by organism of apoptotic process in other organism involved in symbiotic interaction
oxidation-reduction process
antimicrobial humoral immune response mediated by antimicrobial peptide
cellular response to interferon-gamma
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
microtubule binding
aspartic-type endopeptidase inhibitor activity
peptidyl-cysteine S-nitrosylase activity
identical protein binding
NADP binding
NAD binding
disordered domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0