This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03346023_m1
See other LIMS1 GE Assays ›
Gene Symbol:
LIMS1
Gene Name:
LIM zinc finger domain containing 1
Gene Aliases:
PINCH1
Chromosome Location:
Chr.1: 137514681 - 137578645 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03346023_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIMS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001766.1 NP_001001766.1 2 - 3 222 112
XM_015277838.2 XP_015133324.2 3 - 4 515 112
XM_025147122.1 XP_025002890.1 3 - 4 828 112
XM_025147130.1 XP_025002898.1 3 - 4 509 112
XM_025147132.1 XP_025002900.1 3 - 4 523 112
XM_025147133.1 XP_025002901.1 3 - 4 344 112
XM_025147136.1 XP_025002904.1 3 - 4 374 112
XM_025147138.1 XP_025002906.1 3 - 4 385 112
XM_025147140.1 XP_025002908.1 3 - 4 339 112
XM_025147143.1 XP_025002911.1 3 - 4 303 112
GenBank mRNA AJ719853.1 - 3 - 4 326 112
AY612853.1 - 2 - 3 222 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cell junction protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0