This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03345064_s1
See other C5H14ORF169 GE Assays ›
Gene Symbol:
C5H14ORF169
Gene Name:
chromosome 5 open reading frame, human C14orf169
Gene Aliases:
C14orf169, NO66, RIOX1
Chromosome Location:
Chr.5: 26420929 - 26422805 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03345064_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C5H14ORF169
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031196.1 NP_001026367.1 1 1745 146
GenBank mRNA AJ719363.1 - 1 1745 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

histone H3-K4 demethylation
negative regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
histone demethylase activity (H3-K4 specific)
histone demethylase activity (H3-K36 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0