This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03343431_m1
See other HSPD1 GE Assays ›
Gene Symbol:
HSPD1
Gene Name:
heat shock protein family D (Hsp60) member 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 10221920 - 10231186 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03343431_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSPD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012916.2 NP_001012934.1 3 - 4 522 84
GenBank mRNA AJ719862.1 - 3 - 4 524 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

B cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
positive regulation of T cell mediated immune response to tumor cell
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
apoptotic mitochondrial changes
response to cold
positive regulation of interferon-alpha production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
mitochondrial unfolded protein response
protein refolding
B cell proliferation
T cell activation
positive regulation of macrophage activation
negative regulation of apoptotic process
protein import into mitochondrial intermembrane space
isotype switching to IgG isotypes
protein stabilization
positive regulation of T cell activation
interaction with symbiont
cellular response to interleukin-7
lipopolysaccharide binding
p53 binding
double-stranded RNA binding
protein binding
ATP binding
high-density lipoprotein particle binding
isomerase activity
ubiquitin protein ligase binding
apolipoprotein A-I binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header