This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03341985_m1
See other LSM14A GE Assays ›
Gene Symbol:
LSM14A
Gene Name:
LSM14A, mRNA processing body assembly factor
Gene Aliases:
-
Chromosome Location:
Chr.11: 10872989 - 10891277 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03341985_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LSM14A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012778.1 NP_001012796.1 4 - 5 794 80
XM_015292441.2 XP_015147927.1 4 - 5 813 80
XM_015292442.2 XP_015147928.1 4 - 5 816 80
XM_015292443.2 XP_015147929.1 4 - 5 808 80
GenBank mRNA AJ719907.1 - 4 - 5 794 80
MH331646.1 - 4 - 5 539 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0