This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03341230_m1
See other DNAJB6 GE Assays ›
Gene Symbol:
DNAJB6
Gene Name:
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases:
-
Chromosome Location:
Chr.2: 8738313 - 8788235 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03341230_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNAJB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012556.1 NP_001012574.1 2 - 3 201 75
XM_015281335.2 XP_015136821.1 3 - 4 353 75
XM_025147162.1 XP_025002930.1 2 - 3 212 75
XM_025147163.1 XP_025002931.1 2 - 3 264 75
GenBank mRNA AJ851505.1 - 2 - 3 201 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
actin cytoskeleton organization
extracellular matrix organization
positive regulation of ATPase activity
regulation of protein localization
protein localization to nucleus
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
intermediate filament organization
negative regulation of transcription, DNA-templated
negative regulation of inclusion body assembly
ATPase activator activity
DNA binding
heat shock protein binding
unfolded protein binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0