This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338305_m1
See other C4BPM GE Assays ›
Gene Symbol:
C4BPM
Gene Name:
complement component 4 binding protein, membrane
Gene Aliases:
C4BPA, CREM, Cremp
Chromosome Location:
Chr.26: 2687155 - 2722974 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338305_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C4BPM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204664.2 NP_989995.1 443 169
XM_015298885.2 XP_015154371.1 479 169
XM_015298886.2 XP_015154372.1 479 169
XM_015298887.2 XP_015154373.1 479 169
GenBank mRNA AB035592.1 - 444 169
CR382436.1 - 466 169

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0