This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338081_m1
See other ACTR6 GE Assays ›
Gene Symbol:
ACTR6
Gene Name:
ARP6 actin-related protein 6 homolog
Gene Aliases:
GARPX
Chromosome Location:
Chr.1: 47242488 - 47251479 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338081_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204637.1 NP_989968.1 1 - 2 191 72
GenBank mRNA AB038230.1 - 1 - 2 191 72
AJ719762.1 - 1 - 2 192 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

histone exchange
protein binding
nucleosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0