This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03335313_s1
See other DYL1 GE Assays ›
Gene Symbol:
DYL1
Gene Name:
dynein light chain 1 cytoplasmic
Gene Aliases:
DYNLL1
Chromosome Location:
Chr.15: 9563292 - 9565826 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03335313_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DYL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277879.1 NP_001264808.1 1 50 123
GenBank mRNA CR385793.1 - 1 50 123

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001277879.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

microtubule or microtubule-binding cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0