This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03333645_m1
See other C9ORF58 GE Assays ›
Gene Symbol:
C9ORF58
Gene Name:
allograft inflammatory factor 1-like
Gene Aliases:
AIF1L
Chromosome Location:
Chr.17: 6378639 - 6390714 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03333645_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C9ORF58
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_415461.6 XP_415461.1 2 - 3 179 68
GenBank mRNA CR523365.1 - 2 - 3 138 68
KJ101611.1 - 2 - 3 144 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0