This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03332705_m1
See other APC GE Assays ›
Gene Symbol:
APC
Gene Name:
APC, WNT signaling pathway regulator
Gene Aliases:
APC1, CTC-554D6.1
Chromosome Location:
Chr.Z: 46106966 - 46200283 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03332705_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001233410.5 XP_001233411.3 11 - 12 1540 70
XM_004949280.3 XP_004949337.1 12 - 13 1878 70
XM_004949281.3 XP_004949338.1 12 - 13 1854 70
XM_004949282.3 XP_004949339.1 11 - 12 1842 70
XM_004949283.3 XP_004949340.1 11 - 12 1818 70
XM_004949285.3 XP_004949342.1 12 - 13 1878 70
XM_004949286.3 XP_004949343.1 10 - 11 1463 70
XM_015280443.2 XP_015135929.1 12 - 13 1558 70
XM_015280444.2 XP_015135930.1 12 - 13 1576 70
XM_015280445.1 XP_015135931.1 12 - 13 1509 70
XM_015280446.2 XP_015135932.1 11 - 12 1795 70
XM_015280448.2 XP_015135934.1 11 - 12 1771 70
XM_015280450.2 XP_015135936.1 11 - 12 1665 70
XM_025144933.1 XP_025000701.1 10 - 11 1734 70
GenBank mRNA AJ971234.1 - 6 - 7 821 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0