Assay Details
Gene Symbol: HSPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414655.6 | XP_414655.2 | 12 - 13 | 2696 | 65 | |
GenBank mRNA | BX931422.1 | - | 4 - 5 | 583 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
