This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03331072_s1
See other GPR160 GE Assays ›
Gene Symbol:
GPR160
Gene Name:
G protein-coupled receptor 160
Gene Aliases:
-
Chromosome Location:
Chr.9: 20022138 - 20025627 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03331072_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR160
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277472.1 NP_001264401.1 1 1608 149
XM_015291665.2 XP_015147151.1 2 1965 151
GenBank mRNA BX929715.1 - 1 1047 149
BX950382.1 - 1 1608 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0