This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03329524_m1
See other CYBC1 GE Assays ›
Gene Symbol:
CYBC1
Gene Name:
cytochrome b-245 chaperone 1
Gene Aliases:
C17orf62, C18H17orf62
Chromosome Location:
Chr.18: 3496726 - 3506905 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03329524_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYBC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_415607.6 XP_415607.1 1 - 2 203 85
GenBank mRNA BX933442.2 - 1 - 2 76 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_415607.6 transcripts.

Set Membership:

Amplicon length between 71 and 85 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0