This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03324710_m1
See other RPL18A GE Assays ›
Gene Symbol:
RPL18A
Gene Name:
ribosomal protein L18a
Gene Aliases:
-
Chromosome Location:
Chr.1: 33723878 - 33726658 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03324710_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL18A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015282735.2 XP_015138221.1 3 - 4 362 118
GenBank mRNA BX933861.1 - 3 - 4 353 118
CR389461.1 - 3 - 4 363 118
CR390345.1 - 3 - 4 342 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0