This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03320993_m1
See other SNRPB2 GE Assays ›
Gene Symbol:
SNRPB2
Gene Name:
small nuclear ribonucleoprotein polypeptide B2
Gene Aliases:
-
Chromosome Location:
Chr.3: 5652599 - 5658851 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03320993_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNRPB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_419331.6 XP_419331.1 3 - 4 450 92
GenBank mRNA BX931436.2 - 3 - 4 431 92
BX932844.2 - 3 - 4 432 92
BX933982.1 - 3 - 4 441 92
BX934618.1 - 3 - 4 444 92
BX935753.1 - 3 - 4 438 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0