This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03316751_m1
See other PRDX1 GE Assays ›
Gene Symbol:
PRDX1
Gene Name:
peroxiredoxin 1
Gene Aliases:
Peroxiredoxin-1
Chromosome Location:
Chr.8: 21501345 - 21507655 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03316751_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRDX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271932.1 NP_001258861.1 5 - 6 65
GenBank mRNA BU221306.1 - 5 - 6 65
BX930147.1 - 5 - 6 458 65
CR353582.1 - 5 - 6 558 65
CR406137.1 - 5 - 6 595 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
cell proliferation
removal of superoxide radicals
natural killer cell activation
regulation of stress-activated MAPK cascade
cellular response to stress
erythrocyte homeostasis
natural killer cell mediated cytotoxicity
hydrogen peroxide catabolic process
leukocyte activation
cell redox homeostasis
oxidation-reduction process
regulation of NIK/NF-kappaB signaling
protein binding
thioredoxin peroxidase activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0