This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06984810_m1
See other CFAP410 GE Assays ›
Gene Symbol:
CFAP410
Gene Name:
cilia and flagella associated protein 410
Gene Aliases:
C26H21orf2
Chromosome Location:
Chr.26: 40845002 - 40857851 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06984810_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFAP410
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001490381.4 XP_001490431.2 461 76
XM_014736418.2 XP_014591904.2 463 76
XM_023630275.1 XP_023486043.1 445 76
XM_023630276.1 XP_023486044.1 443 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0