This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06959968_m1
See other KCNMB3 GE Assays ›
Gene Symbol:
KCNMB3
Gene Name:
potassium calcium-activated channel subfamily M regulatory beta subunit 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 20722790 - 20770559 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06959968_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNMB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014732762.2 XP_014588248.1 16122 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus

Panther Classification:

Molecular Function -

voltage-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0