This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04947721_m1
See other ELOA GE Assays ›
Gene Symbol:
ELOA ELOA
Gene Name:
elongin A
elongin A
Gene Aliases:
TCEB3
-
Chromosome Location:
Chr.2: 31283856 - 31296656 on Build EquCab3.0
Chromosome Location:
Chr.2: 31349923 - 31365249 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
-
Assay ID Ec04947721_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252407.2 NP_001239336.1 2 - 3 149 77
Gene Symbol: ELOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023635511.1 XP_023491279.1 214 77
XM_023635512.1 XP_023491280.1 227 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Equus caballus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0