Assay Details
Gene Symbol: MIR9110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128139.1 | 1 | 83 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100072669 | LOC100072669 | P2RX5 | Sense | Chr. 11 - 47559680 - 47586088 on NCBI Build EquCab3.0 | |
Downstream | integrin subunit alpha E | ITGAE | 27.3kb | Sense | Chr. 11 - 47591040 - 47660437 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
