Assay Details
Gene Symbol: MIR9076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128099.1 | 1 | 82 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinase insert domain receptor | KDR | FLK | -65.2kb | Anti-sense | Chr. 3 - 79241286 - 79287292 on NCBI Build EquCab3.0 |
Downstream | folate receptor alpha-like | LOC100059683 | 8.0kb | Anti-sense | Chr. 3 - 79360546 - 79361618 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
