Assay Details
Gene Symbol: MIR670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032923.1 | 1 | 57 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tetratricopeptide repeat domain 17 | TTC17 | -49.4kb | Sense | Chr. 12 - 8377104 - 8492147 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-129b | MIR129B | eca-mir-129b | 21.6kb | Sense | Chr. 12 - 8563202 - 8563275 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
