This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec03468715_m1
See other HAPLN1 GE Assays ›
Gene Symbol:
HAPLN1
Gene Name:
hyaluronan and proteoglycan link protein 1
Gene Aliases:
CTRL1
Chromosome Location:
Chr.14: 83172580 - 83246397 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec03468715_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAPLN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082504.1 NP_001075973.1 1 - 2 321 82
GenBank mRNA X78077.1 - 321 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001082504.1, X78077.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Equus caballus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0