This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03439763_g1
See other KHDRBS1B GE Assays ›
Gene Symbol:
khdrbs1b
Gene Name:
KH domain containing, RNA binding, signal transduction associated 1b
Gene Aliases:
khdrbs1l, sb:cb97, wu:fb07d11, wu:fi43c05, zgc:85948
Chromosome Location:
Chr.19: 30347509 - 30359077 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03439763_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: khdrbs1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213235.1 NP_998400.1 3 - 4 486 90
GenBank mRNA BC067711.1 - 3 - 4 486 90
BC155179.1 - 3 - 4 507 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
nucleic acid binding
RNA binding
mRNA binding
poly(A) binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0