This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03438681_m1
See other PITX3 GE Assays ›
Gene Symbol:
pitx3
Gene Name:
paired-like homeodomain 3
Gene Aliases:
zgc:109716
Chromosome Location:
Chr.13: 7556314 - 7573708 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03438681_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pitx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205675.2 NP_991238.1 2 - 3 343 73
GenBank mRNA AY525643.1 - 2 - 3 347 73
AY639155.1 - 1 - 2 134 73
BC094961.1 - 2 - 3 328 73
BC165321.1 - 1 - 2 150 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
regulation of transcription, DNA-templated
multicellular organism development
anatomical structure morphogenesis
adenohypophysis development
endocrine system development
camera-type eye development
positive regulation of transcription, DNA-templated
lens fiber cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0