This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03437053_m1
See other RBM14A GE Assays ›
Gene Symbol:
rbm14a
Gene Name:
RNA binding motif protein 14a
Gene Aliases:
im:7053198, wu:fc08a04, zgc:110682
Chromosome Location:
Chr.21: 27370640 - 27382659 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03437053_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbm14a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001115144.1 NP_001108616.1 3 - 4 1210 179
XM_021468755.1 XP_021324430.1 3 - 4 1078 179
GenBank mRNA BC095368.1 - 3 - 4 1210 179
BC157376.1 - 3 - 4 1210 179

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Process(es)

nucleic acid binding
RNA binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0