This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434035_m1
See other MIER1A GE Assays ›
Gene Symbol:
mier1a
Gene Name:
mesoderm induction early response 1a, transcriptional regulator
Gene Aliases:
mier1, wu:fb30a02, zgc:56179
Chromosome Location:
Chr.6: 34825749 - 34845462 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434035_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mier1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199559.1 NP_955853.1 9 - 10 885 148
XM_017356435.2 XP_017211924.1 11 - 12 1132 148
XM_017356436.2 XP_017211925.1 11 - 12 1129 148
XM_021476778.1 XP_021332453.1 11 - 12 1136 148
XM_021476779.1 XP_021332454.1 11 - 12 1133 148
GenBank mRNA BC045973.1 - 9 - 10 885 148
BC164544.1 - 8 - 9 845 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
histone deacetylation
RNA polymerase II repressing transcription factor binding
DNA binding
transcription corepressor activity
histone deacetylase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0