This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434033_g1
See other MIER1A GE Assays ›
Gene Symbol:
mier1a
Gene Name:
mesoderm induction early response 1a, transcriptional regulator
Gene Aliases:
mier1, wu:fb30a02, zgc:56179
Chromosome Location:
Chr.6: 34825749 - 34845462 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434033_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mier1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199559.1 NP_955853.1 7 - 8 649 91
XM_017356435.2 XP_017211924.1 9 - 10 896 91
XM_017356436.2 XP_017211925.1 9 - 10 893 91
XM_021476778.1 XP_021332453.1 9 - 10 900 91
XM_021476779.1 XP_021332454.1 9 - 10 897 91
GenBank mRNA BC045973.1 - 7 - 8 649 91
BC164544.1 - 6 - 7 609 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
histone deacetylation
RNA polymerase II repressing transcription factor binding
DNA binding
transcription corepressor activity
histone deacetylase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0