This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03433980_m1
See other MNS1 GE Assays ›
Gene Symbol:
mns1
Gene Name:
meiosis-specific nuclear structural 1
Gene Aliases:
sb:cb494, zgc:65845
Chromosome Location:
Chr.7: 81408 - 90124 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03433980_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mns1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199433.1 NP_955465.1 7 - 8 1120 55
XM_009303233.3 XP_009301508.1 7 - 8 1120 55
XM_021477594.1 XP_021333269.1 11 - 12 1683 55
GenBank mRNA BC059798.1 - 7 - 8 1120 55
BC152207.1 - 7 - 8 1088 55
BC171355.1 - 7 - 8 1060 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s) Process(es)

cilium organization
meiotic cell cycle
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0