This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03432086_g1
See other ABLIM1A GE Assays ›
Gene Symbol:
ablim1a
Gene Name:
actin binding LIM protein 1a
Gene Aliases:
ablim1, zgc:172321
Chromosome Location:
Chr.13: 43507383 - 43599920 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03432086_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ablim1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114316.1 NP_001107788.1 16 - 17 1599 92
XM_009307552.3 XP_009305827.1 17 - 18 4187 92
XM_009307554.3 XP_009305829.1 17 - 18 4160 92
XM_009307559.3 XP_009305834.1 16 - 17 4040 92
XM_009307565.3 XP_009305840.1 17 - 18 4154 92
XM_017358665.2 XP_017214154.1 16 - 17 4076 92
XM_017358666.2 XP_017214155.1 16 - 17 4070 92
XM_017358667.2 XP_017214156.1 15 - 16 3928 92
XM_021480476.1 XP_021336151.1 18 - 19 4233 92
XM_021480477.1 XP_021336152.1 18 - 19 4215 92
XM_021480478.1 XP_021336153.1 18 - 19 4205 92
XM_021480479.1 XP_021336154.1 17 - 18 4130 92
XM_021480480.1 XP_021336155.1 17 - 18 4085 92
XM_021480481.1 XP_021336156.1 17 - 18 4079 92
XM_021480482.1 XP_021336157.1 16 - 17 4001 92
XM_021480483.1 XP_021336158.1 16 - 17 3995 92
GenBank mRNA BC155652.1 - 16 - 17 1599 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
lamellipodium assembly
cilium morphogenesis
actin binding
metal ion binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0