This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03428918_g1
See other EIF4G1A GE Assays ›
Gene Symbol:
eif4g1a
Gene Name:
eukaryotic translation initiation factor 4 gamma, 1a
Gene Aliases:
CH73-211A15.2, fb50a05, wu:fb50a05, wu:fc60b06, zgc:158450
Chromosome Location:
Chr.2: 16798610 - 16839132 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03428918_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif4g1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080200.1 NP_001073669.1 12 - 13 1954 143
XM_009298328.3 XP_009296603.1 15 - 16 2162 143
GenBank mRNA BC129235.1 - 12 - 13 1954 143

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Process(es)

RNA binding
mRNA binding
translation initiation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0