This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03426747_g1
See other ID4 GE Assays ›
Gene Symbol:
id4
Gene Name:
inhibitor of DNA binding 4
Gene Aliases:
cb547, fb77d04, sb:cb547, wu:fb77d04, zgc:123214
Chromosome Location:
Chr.16: 9977858 - 9980799 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03426747_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: id4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039990.1 NP_001035079.1 1 - 2 499 90
GenBank mRNA BC081684.1 - 1 - 2 487 90
BC093226.1 - 1 - 2 499 90
BC108045.1 - 1 - 2 499 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
atrioventricular valve development
regulation of Wnt signaling pathway involved in heart development
cell differentiation
circadian regulation of gene expression
atrioventricular canal development
negative regulation of transcription, DNA-templated
protein dimerization activity
transcription regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0