This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03424964_g1
See other HERC5.3 GE Assays ›
Gene Symbol:
herc5.3 herc5.4
Gene Name:
HECT and RLD domain containing E3 ubiquitin protein ligase 5.3
HECT and RLD domain containing E3 ubiquitin protein ligase 5.4
Gene Aliases:
herc56.3, mate4, slc47a2.2, zgc:163136
her56.4, si:ch211-281g13.5
Chromosome Location:
Chr.1: 49532653 - 49544601 on Build GRCz11
Chromosome Location:
Chr.1: 49524432 - 49530868 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
Assay ID Dr03424964_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: herc5.3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098763.1 NP_001092233.1 9 - 10 1126 133
GenBank mRNA BC139897.1 - 9 - 10 1126 133
Gene Symbol: herc5.4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145631.1 NP_001139103.1 8 - 9 1121 133
XM_005160109.4 XP_005160166.1 8 - 9 1374 133
XM_017355671.2 XP_017211160.1 9 - 10 1281 133
XM_021475845.1 XP_021331520.1 9 - 10 1291 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Process(es)

ubiquitin-protein transferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0