This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03421948_g1
See other RPL9 GE Assays ›
Gene Symbol:
rpl9
Gene Name:
ribosomal protein L9
Gene Aliases:
ab02c03, fa93a01, mg:ab02c03, wu:fa93a01, zgc:103730
Chromosome Location:
Chr.1: 5196 - 9507 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03421948_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rpl9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003861.1 NP_001003861.1 1 - 2 48 65
XM_005170906.4 XP_005170963.1 2 - 3 117 65
XM_005170907.4 XP_005170964.1 2 - 3 109 65
GenBank mRNA AY648736.1 - 1 - 2 48 65
BC090911.1 - 2 - 3 132 65
BC115139.1 - 2 - 3 74 65
BC152564.1 - 2 - 3 70 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC152564.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
translation
nucleotide binding
DNA binding
DNA clamp loader activity
structural constituent of ribosome
ATP binding
rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0