This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03201662_s1
See other PPP2R5EB GE Assays ›
Gene Symbol:
ppp2r5eb
Gene Name:
protein phosphatase 2, regulatory subunit B', epsilon isoform b
Gene Aliases:
ppp2r5e, ppp2r5e1, wdb2
Chromosome Location:
Chr.13: 37421333 - 37465524 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03201662_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppp2r5eb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194412.2 NP_919393.2 1 47 84
GenBank mRNA BC048034.1 - 1 47 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC048034.1, NM_194412.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
signal transduction
regulation of protein autophosphorylation
protein phosphatase regulator activity
protein phosphatase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0