This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03183723_s1
See other RGS3A GE Assays ›
Gene Symbol:
rgs3a
Gene Name:
regulator of G protein signaling 3a
Gene Aliases:
id:ibd5096, rgs3, zgc:100933
Chromosome Location:
Chr.5: 269881 - 299313 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03183723_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003629.1 NP_001003629.1 5 1254 135
XM_005168407.4 XP_005168464.1 7 2287 135
XM_005168409.2 XP_005168466.1 5 1294 135
XM_009302037.3 XP_009300312.2 17 3474 135
XM_009302039.3 XP_009300314.1 8 2300 135
XM_017356067.2 XP_017211556.1 5 1330 135
XM_021476089.1 XP_021331764.1 7 2218 135
GenBank mRNA BC078194.1 - 5 1254 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s)

regulation of Wnt signaling pathway, calcium modulating pathway
negative regulation of Wnt signaling pathway, calcium modulating pathway
somite development

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0