This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03150697_m1
See other TCERG1A GE Assays ›
Gene Symbol:
tcerg1a LOC103910002
Gene Name:
transcription elongation regulator 1a (CA150)
transcription elongation regulator 1-like
Gene Aliases:
tcerg1, wu:fb80g01
-
Chromosome Location:
Chr.14: 52481292 - 52518568 on Build GRCz11
Chromosome Location:
Chr.Unknown: 7388 - 12546 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
Assay ID Dr03150697_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tcerg1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198357.3 NP_938171.2 4 - 5 725 77
GenBank mRNA BC056813.1 - 4 - 5 741 77
Gene Symbol: LOC103910002
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009298386.3 XP_009296661.2 4 - 5 754 77
XM_021471000.1 XP_021326675.1 4 - 5 742 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Process(es)

transcription cofactor activity
RNA polymerase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0