This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03149960_m1
See other TRIOA GE Assays ›
Gene Symbol:
trioa
Gene Name:
trio Rho guanine nucleotide exchange factor a
Gene Aliases:
si:ch211-13c6.1, si:dkey-158b13.2, trio
Chromosome Location:
Chr.19: 41840964 - 41991299 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03149960_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: trioa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001104526.1 NP_001097996.1 4 - 5 540 65
XM_009294421.3 XP_009292696.1 4 - 5 707 65
XM_017352243.2 XP_017207732.1 4 - 5 711 65
XM_021468075.1 XP_021323750.1 4 - 5 707 65
XM_021468076.1 XP_021323751.1 4 - 5 708 65
XM_021468077.1 XP_021323752.1 4 - 5 708 65
XM_021468078.1 XP_021323753.1 4 - 5 709 65
XM_021468079.1 XP_021323754.1 4 - 5 710 65
XM_021468080.1 XP_021323755.1 4 - 5 711 65
XM_021468081.1 XP_021323756.1 4 - 5 711 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
axon guidance
central nervous system development
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0