This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03144492_m1
See other GPIA GE Assays ›
Gene Symbol:
gpia
Gene Name:
glucose-6-phosphate isomerase a
Gene Aliases:
PGI, gpi, pgi-1, wu:fc02c05, zgc:86919
Chromosome Location:
Chr.25: 35913678 - 35938996 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03144492_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gpia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144763.1 NP_658909.1 5 - 6 576 135
GenBank mRNA AJ306395.1 - 5 - 6 576 135
BC044450.1 - 5 - 6 562 135
BC083507.1 - 5 - 6 560 135
BC165764.1 - 5 - 6 517 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
glucose 6-phosphate metabolic process
glucose-6-phosphate isomerase activity
isomerase activity
monosaccharide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0