This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141969_mH
See other DSCAMA GE Assays ›
Gene Symbol:
dscama
Gene Name:
Down syndrome cell adhesion molecule a
Gene Aliases:
dscam
Chromosome Location:
Chr.10: 38775334 - 38959079 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141969_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dscama
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030224.2 NP_001025395.1 7 - 8 2065 73
XM_021479191.1 XP_021334866.1 7 - 8 2142 73
GenBank mRNA AY552547.1 - 7 - 8 2065 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
nervous system development
axon guidance
cell migration involved in gastrulation
dendrite self-avoidance
protein binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0