This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141502_m1
See other DNMT3AB GE Assays ›
Gene Symbol:
dnmt3ab
Gene Name:
DNA (cytosine-5-)-methyltransferase 3 alpha b
Gene Aliases:
dnmt3a1, dnmt6
Chromosome Location:
Chr.17: 36937808 - 37054963 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141502_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnmt3ab
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001018140.2 NP_001018150.1 12 - 13 1855 69
NM_001328167.1 NP_001315096.1 17 - 18 2404 69
XM_005158845.4 XP_005158902.1 12 - 13 1769 69
XM_009293199.3 XP_009291474.1 16 - 17 2291 69
XM_009293200.3 XP_009291475.1 12 - 13 1862 69
XM_021467161.1 XP_021322836.1 17 - 18 2544 69
XM_021467162.1 XP_021322837.1 17 - 18 2454 69
XM_021467163.1 XP_021322838.1 17 - 18 2455 69
GenBank mRNA AB196917.1 - 12 - 13 1857 69
BC162582.1 - 12 - 13 1767 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
regulation of gene expression
methylation
methyltransferase activity
DNA-methyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0