This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03141128_m1
See other FN1B GE Assays ›
Gene Symbol:
fn1b
Gene Name:
fibronectin 1b
Gene Aliases:
cb1057, fn1l, fn3, wu:fa14f11, wu:fb03c02
Chromosome Location:
Chr.1: 5504131 - 5543086 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03141128_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fn1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013261.1 NP_001013279.1 26 - 27 4158 77
XM_005160284.3 XP_005160341.1 26 - 27 4190 77
XM_009305554.3 XP_009303829.1 27 - 28 4470 77
GenBank mRNA AY538257.1 - 26 - 27 4158 77
AY725818.1 - 27 - 28 4449 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

somitogenesis
heart field specification
atrioventricular valve morphogenesis
cell-substrate junction assembly
cell-matrix adhesion
nervous system development
heart development
wound healing
platelet aggregation
blood coagulation, fibrin clot formation
receptor binding
integrin binding
proteoglycan binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0